2-Amino-5-chloropyrazine 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-amino-5-chloropyrazine

H66509-06 H66509-14
AAH66509-03EA 30.94 USD
AAH66509-03 AAH66509-06 AAH66509-14
2-Amino-5-chloropyrazine 95%
2-Amino-5-chloropyrazine

2-Amino-5-chloropyrazine, 95%


Purity: 95% CAS No.: 33332-29-5 Molecular Formula: C4H4ClN3 Formula Weight: 129.55 Storage Temperature: 2°C to 8°C Physical Form: Solid Melting Point: 122-128° Beilstein Reference: 507879 MDL No.: MFCD03423596

Formula: C₄H₄ClN₃
MW: 129.55 g/mol
Melting Pt: 122…128 °C
Storage Temperature: Ambient
MDL Number: MFCD03423596
CAS Number: 33332-29-5

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR