Fmoc-D-allo-thr(tBu)-OH 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Fmoc-O-tert-butyl-D-allo-threonine, Fmoc-D-Allo-Thr(tBu)-OH
H66486-03
H66486-MD
AAH66486-03EA
178
USD
AAH66486-03
AAH66486-MD
Fmoc-D-allo-thr(tBu)-OH 98%
Fmoc-D-allo-thr(tBu)-OH
N-Fmoc-O-tert-butyl-D-allo-threonine, 98%
Purity: 98%
CAS No.: 170643-02-4
Molecular Formula: C23H27NO5
Formula Weight: 397.47
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00077074
Formula:
C₂₃H₂₇NO₅ MW: 397.471 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00077074 CAS Number: 170643-02-4 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...