3-Fluoro-4-hydroxy-5-methoxybenzaldehyde 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Fluoro-4-hydroxy-5-methoxybenzene
H66477-03
H66477-MD
AAH66477-03EA
236
USD
AAH66477-03
AAH66477-MD
3-Fluoro-4-hydroxy-5-methoxybenzaldehyde 96%
3-Fluoro-4-hydroxy-5-methoxybenzaldehyde
3-Fluoro-4-hydroxy-5-methoxybenzaldehyde, 96%
Purity: 96%
CAS No.: 79418-78-3
Molecular Formula: C8H7FO3
Formula Weight: 170.14
Physical Form: Powder
Sensitivity: Air Sensitive
Appearance: Yelloe to pale brown
Boiling Point: 114-118°
MDL No.: MFCD02683560
Formula:
C₈H₇FO₃ MW: 170.139 g/mol Boiling Pt: 114…118 °C Storage Temperature: Ambient |
MDL Number:
MFCD02683560 CAS Number: 79418-78-3 |
Specification Test Results
Assay Minimum | 96 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...