2-Chloro-6-(trichloromethyl)pyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Nitrapyrin
H66418-03
H66418-06
AAH66418-03EA
37.1
USD
AAH66418-03
AAH66418-06
2-Chloro-6-(trichloromethyl)pyridine 98%
2-Chloro-6-(trichloromethyl)pyridine
2-Chloro-6-(trichloromethyl)pyridine, 98%
Purity: 98%
CAS No.: 1929-82-4
Molecular Formula: C6H3Cl4N
Formula Weight: 230.90
Flash Point: 100°(212°F)
Physical Form: Powder
Appearance: White
Melting Point: 60-65°
Beilstein Reference: 1618997
UN No.: 3077
MDL No.: MFCD00072500
Formula:
C₆H₃Cl₄N MW: 230.897 g/mol Melting Pt: 60…65 °C Flash Pt: 100 °C(212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00072500 CAS Number: 1929-82-4 EINECS: 217-682-2 UN: 3077 ADR: "9",III |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...