4-Phenylpiperidine 96%

Supplier: Thermo Scientific Chemicals
Warning

H66389-03 H66389-06
AAH66389-03EA 43.4 USD
AAH66389-03 AAH66389-06
4-Phenylpiperidine 96%
4-Phenylpiperidine

4-Phenylpiperidine, 96%


Purity: 96% CAS No.: 771-99-3 Molecular Formula: C11H15N Formula Weight: 161.24 Physical Form: Powder Appearance: Yellow to brown Melting Point: 61-64° Beilstein Reference: 124508 MDL No.: MFCD00006002

Formula: C₁₁H₁₅N
MW: 161.25 g/mol
Melting Pt: 61…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00006002
CAS Number: 771-99-3
EINECS: 212-243-1

Order Now

Specification Test Results

Assay Minimum 96

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR