Fmoc-Glu(OAll)-OH 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Fmoc-L-glutamic acid 5-allyl ester, N-Fmoc-L-glutamic acid 5-Allyl ester, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamic acid 5-Allyl ester, Fmoc-Glu(allyl ester)-OH, 5-Allyl N-Fmoc-L-glutamate, Fmoc-Glu(OAll)-OH, 5-Allyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamate
N-Fmoc-L-glutamic acid 5-allyl ester, 95%
Purity: 95%
CAS No.: 133464-46-7
Molecular Formula: C23H23NO6
Formula Weight: 409.44
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Beilstein Reference: 6670846
Optical Rotation: -16°(c=1 in DMF)
MDL No.: MFCD00190879
Formula:
C₂₃H₂₃NO₆ MW: 409.438 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00190879 CAS Number: 133464-46-7 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...