3,5-Dimethyl-1-adamantaneacetic acid 97%
Supplier: Thermo Scientific Chemicals
3,5-Dimethyl-1-adamantaneacetic acid, 97%
Purity: 97%
CAS No.: 14202-14-3
Molecular Formula: C14H22O2
Formula Weight: 222.32
Physical Form: Powder
Appearance: White to off-white
Melting Point: 110-115°
MDL No.: MFCD00077207
Formula:
C₁₄H₂₂O₂ Melting Pt: 110…115 °C Storage Temperature: Ambient |
MDL Number:
MFCD00077207 CAS Number: 14202-14-3 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...