4-[[(9H-Fluoren-9-ylmethoxy)carbonyl]aminomethyl]benzoic acid 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Fmoc-4-Amb-OH, 4-(Fmoc-aminomethyl)benzoic Acid

H66340-06 H66340-14
AAH66340-06EA 69.7 USD
AAH66340-06 AAH66340-14
4-[[(9H-Fluoren-9-ylmethoxy)carbonyl]aminomethyl]benzoic acid 95%
4-[[(9H-Fluoren-9-ylmethoxy)carbonyl]aminomethyl]benzoic acid

4-(Fmoc-aminomethyl)benzoic acid, 95%


Purity: 95% CAS No.: 164470-64-8 Molecular Formula: C23H19NO4 Formula Weight: 373.41 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Melting Point: 224-228° Beilstein Reference: 7391207 MDL No.: MFCD01074693

Formula: C₂₃H₁₉NO₄
MW: 373.41 g/mol
Melting Pt: 224…228 °C
Storage Temperature: Ambient
MDL Number: MFCD01074693
CAS Number: 164470-64-8

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR