2-Methyl-5-pyridineboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(6-Methylpyridin-3-yl)boronic acid
H66327-03
H66327-06
AAH66327-03EA
122
USD
AAH66327-03
AAH66327-06
AAH66327-MD
2-Methyl-5-pyridineboronic acid 97%
2-Methyl-5-pyridineboronic acid
2-Methylpyridine-5-boronic acid, 97%
Purity: 97%
CAS No.: 659742-21-9
Molecular Formula: C6H8BNO2
Formula Weight: 136.94
Physical Form: Powder
Appearance: White to pale brown
MDL No.: MFCD06801736
Formula:
C₆H₈BNO₂ MW: 136.94 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06801736 CAS Number: 659742-21-9 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...