5-Iodo-2-methoxybenzaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66320-03
H66320-06
AAH66320-03EA
74.8
USD
AAH66320-03
AAH66320-06
5-Iodo-2-methoxybenzaldehyde 97%
5-Iodo-2-methoxybenzaldehyde
5-Iodo-2-methoxybenzaldehyde, 97%
Purity: 97%
CAS No.: 42298-41-9
Molecular Formula: C8H7IO2
Formula Weight: 262.04
Physical Form: Solid
Sensitivity: Air and Light Sensitive
Appearance: White to off-white to yel
Melting Point: 140-146°
MDL No.: MFCD04128999
Formula:
C₈H₇IO₂ Melting Pt: 140…146 °C Storage Temperature: Ambient |
MDL Number:
MFCD04128999 CAS Number: 42298-41-9 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...