N-Fmoc-8-aminooctanoic acid 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Fmoc-8-Aoc-OH, Fmoc-8-aminooctanoic acid

H66279-03 H66279-06
AAH66279-03EA 120 USD
AAH66279-03 AAH66279-06
N-Fmoc-8-aminooctanoic acid 95%
N-Fmoc-8-aminooctanoic acid

8-(Fmoc-amino)octanoic acid, 95%


Purity: 95% CAS No.: 126631-93-4 Molecular Formula: C23H27NO4 Formula Weight: 381.47 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Beilstein Reference: 8286497 MDL No.: MFCD00235890

Formula: C₂₃H₂₇NO₄
MW: 381.472 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00235890
CAS Number: 126631-93-4

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR