((3-Bromopropoxy)methyl)benzene 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzyl 3-bromopropyl ether, 3-bromopropylbenzyl ether

H66278-03 H66278-06
AAH66278-03EA 37.1 USD
AAH66278-03 AAH66278-06
((3-Bromopropoxy)methyl)benzene 95%
((3-Bromopropoxy)methyl)benzene

Benzyl 3-bromopropyl ether, 95%


Purity: 95% CAS No.: 54314-84-0 Molecular Formula: C10H13BrO Formula Weight: 229.12 Flash Point: 113°(235°F) Physical Form: Liquid Density: 1.298 Refractive Index: 1.531 Appearance: Colorless Boiling Point: 130-132°/8mm Beilstein Reference: 2084412 MDL No.: MFCD00134570

Formula: C₁₀H₁₃BrO
Boiling Pt: 130…132 °C/8 mm
Density: 1.298
Flash Pt: 113 °C(235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00134570
CAS Number: 54314-84-0

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR