2-(Bromomethyl)oxetane 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66276-03
H66276-MD
AAH66276-03EA
1007
USD
AAH66276-03
AAH66276-MD
2-(Bromomethyl)oxetane 96%
2-(Bromomethyl)oxetane
2-(Bromomethyl)oxetane, 96%
Purity: 96%
CAS No.: 939759-23-6
Molecular Formula: C4H7BrO
Formula Weight: 151.00
Physical Form: Liquid
Sensitivity: Moisture Sensitive
Appearance: Colorless to pale yellow
MDL No.: MFCD08544405
Formula:
C₄H₇BrO Storage Temperature: Ambient |
MDL Number:
MFCD08544405 CAS Number: 939759-23-6 |
Specification Test Results
Assay Minimum | 96 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...