4-(Fmoc-aminomethyl)cyclohexane-1-carboxylic acid 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Fmoc-4-Amc-OH
H66266-03
H66266-06
AAH66266-03EA
85.1
USD
AAH66266-03
AAH66266-06
AAH66266-MD
4-(Fmoc-aminomethyl)cyclohexane-1-carboxylic acid 98%
4-(Fmoc-aminomethyl)cyclohexane-1-carboxylic acid
4-(Fmoc-aminomethyl)cyclohexane-1-carboxylic acid, 98%
Purity: 98%
CAS No.: 188715-40-4
Molecular Formula: C23H25NO4
Formula Weight: 379.18
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: Off-white to white
Beilstein Reference: 7831105
MDL No.: MFCD01862347
Formula:
C₂₃H₂₅NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD01862347 CAS Number: 188715-40-4 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...