5-(Fmoc-amino)valeric acid 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Fmoc-5-aminopentanoic acid, Fmoc-5-Ava-OH
H66264-03
H66264-06
H66264-14
AAH66264-03EA
46.2
USD
AAH66264-03
AAH66264-06
AAH66264-14
5-(Fmoc-amino)valeric acid 98%
5-(Fmoc-amino)valeric acid
5-(Fmoc-amino)valeric acid, 98%
Purity: 98%
CAS No.: 123622-48-0
Molecular Formula: C20H21NO4
Formula Weight: 339.39
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Beilstein Reference: 3565625
MDL No.: MFCD00235889
Formula:
C₂₀H₂₁NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00235889 CAS Number: 123622-48-0 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...