3-Chloroanthranilic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
2-Amino-3-chlorobenzoic acid, 98%
Purity: 98%
CAS No.: 6388-47-2
Molecular Formula: C7H6ClNO2
Formula Weight: 171.58
Physical Form: Solid
Sensitivity: Air and Light Sensitive
Appearance: Yellow to brown
Melting Point: 189-193°
Beilstein Reference: 2090431
MDL No.: MFCD00134229
Formula:
C₇H₆ClNO₂ Melting Pt: 189…193 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134229 CAS Number: 6388-47-2 EINECS: 228-996-4 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...