N-Cbz-N'-boc-L-ornithine 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (S)-2-(((Benzyloxy)carbonyl)amino)-5-((tert-butoxycarbonyl)amino)pentanoic acid, Nα-Benzyloxycarbonyl-Nδ-Boc-L-ornithine, Z-Orn(Boc)-OH

H66247-06 H66247-14
AAH66247-06EA 94.5 USD
AAH66247-06 AAH66247-14
N-Cbz-N'-boc-L-ornithine 98%
N-Cbz-N'-boc-L-ornithine

N^a-Benzyloxycarbonyl-N^d-Boc-L-ornithine, 98%


Purity: 98% CAS No.: 7733-29-1 Molecular Formula: C18H26N2O6 Formula Weight: 366.41 Physical Form: Solid Melting Point: 97-103° Beilstein Reference: 3086976 MDL No.: MFCD00276410

Formula: C₁₈H₂₆N₂O₆
Melting Pt: 97…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00038268
CAS Number: 7733-29-1

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR