2-Hydroxymethyloxetane 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Oxetanemethanol, Oxetan-2-ylmethanol, 2,4-Epoxybutanol

H66218-03 H66218-MD
AAH66218-03EA 358 USD
AAH66218-03 AAH66218-MD
2-Hydroxymethyloxetane 96%
2-Hydroxymethyloxetane

2-Oxetanemethanol, 96%


Purity: 96% CAS No.: 61266-70-4 Molecular Formula: C4H8O2 Formula Weight: 88.11 Storage Temperature: -30°C to -10°C Physical Form: Liquid Density: 1.080 Refractive Index: 1.4460 Sensitivity: Moisture Sensitive Appearance: Colorless to yellow Boiling Point: 98°/15mm MDL No.: MFCD00142933

Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 98 °C/15 mm
Density: 1.080
Storage Temperature: Refrigerator
MDL Number: MFCD00142933
CAS Number: 61266-70-4

Order Now

Specification Test Results

Assay Minimum 96

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR