4-Aminoimidazole-5-carboxamide 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Amino-1H-imidazole-5-carboxamide, 5-Aminoimidazole-4-carboxamide, 5-Amino-4-imidazolecarboxamide, 5-Amino-1H-imidazole-4-carboxamide, 4-Aminoimidazole-5-carboxamide, 4-aminoimidazole-5-carboxamide, 5-Amino-3H-imidazole-4-Carboxamide
H66184-06
H66184-14
AAH66184-06EA
43.4
USD
AAH66184-06
AAH66184-14
AAH66184-22
4-Aminoimidazole-5-carboxamide 95%
4-Aminoimidazole-5-carboxamide
4-Aminoimidazole-5-carboxamide, 95%
Purity: 95%
CAS No.: 360-97-4
Molecular Formula: C4H6N4O
Formula Weight: 126.12
Physical Form: Powder
Appearance: White
Melting Point: 164-170°
Beilstein Reference: 3938
MDL No.: MFCD02181040
Formula:
C4H6N4O MW: 126.12 g/mol Melting Pt: 164…170 °C Storage Temperature: Ambient |
MDL Number:
MFCD02181040 CAS Number: 360-97-4 EINECS: 206-641-4 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...