Fmoc-GABA-OH 95%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-(Fmoc-amino)butyric acid, N-Fmoc-4-aminobutyric acid, Fmoc-γ-Abu-OH, Fmoc-GABA-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-aminobutyric acid

H66147-06 H66147-14
AAH66147-06EA 80.1 USD
AAH66147-06 AAH66147-14
Fmoc-GABA-OH 95%
Fmoc-GABA-OH

4-(Fmoc-amino)butyric acid, 95%


Purity: 95% CAS No.: 116821-47-7 Molecular Formula: C19H19NO4 Formula Weight: 325.36 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Beilstein Reference: 7491485 MDL No.: MFCD00144889

Formula: C₁₉H₁₉NO₄
MW: 325.364 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00144889
CAS Number: 116821-47-7

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR