4-Bromoresorcinol 98%
Supplier: Thermo Scientific Chemicals
4-Bromoresorcinol, 98%
Purity: 98%
CAS No.: 6626-15-9
Molecular Formula: C6H5BrO2
Formula Weight: 189.01
Physical Form: Powder
Appearance: Pink
Odor: Odorless
Melting Point: 97-100°
Boiling Point: 150°/12mm
Beilstein Reference: 1861280
MDL No.: MFCD00002272
Formula:
C₆H₅BrO₂ Boiling Pt: 150 °C/12 mm Melting Pt: 97…100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002272 CAS Number: 6626-15-9 EINECS: 229-586-8 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...