1-Benzyloxycarbonyl-4-oxo-L-proline 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(S)-1-((Benzyloxy)carbonyl)-4-oxopyrrolidine-2-carboxylic acid
H66054-03
H66054-06
AAH66054-03EA
127
USD
AAH66054-03
AAH66054-06
AAH66054-MD
1-Benzyloxycarbonyl-4-oxo-L-proline 97%
1-Benzyloxycarbonyl-4-oxo-L-proline
1-Benzyloxycarbonyl-4-oxo-L-proline, 97%
Purity: 97%
CAS No.: 64187-47-9
Molecular Formula: C13H13NO5
Formula Weight: 263.24
Physical Form: Powder
Appearance: White to yellow to brown
Melting Point: 95-100°
Optical Rotation: +6.0° (c=1 in chloroform)
MDL No.: MFCD07367987
Formula:
C₁₃H₁₃NO₅ MW: 263.249 g/mol Melting Pt: 90…95 °C Storage Temperature: Ambient |
MDL Number:
MFCD07367987 CAS Number: 64187-47-9 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...