L-β-Homoleucine hydrochloride 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
H-β-HomoLeu-OH·HCl
H65857-03
H65857-MD
AAH65857-03EA
394
USD
AAH65857-03
AAH65857-MD
L-β-Homoleucine hydrochloride 95%
L-beta-Homoleucine hydrochloride
L-^b-Homoleucine hydrochloride, 95%
Purity: 95%
CAS No.: 96386-92-4
Molecular Formula: C7H15NO2.HCl
Formula Weight: 181.66
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to pale brown
Beilstein Reference: 8073252
MDL No.: MFCD01862871
Formula:
C₇H₁₅NO₂·HCl MW: 181.66 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01862871 CAS Number: 96386-92-4 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...