N-Methyl-D-alanine 98%
Supplier: Thermo Scientific Chemicals
N-Methyl-D-alanine, 98%
Purity: 98%
CAS No.: 29475-64-7
Molecular Formula: C4H9NO2
Formula Weight: 103.12
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Beilstein Reference: 1720923
MDL No.: MFCD00063135
Formula:
C₄H₉NO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD00063135 CAS Number: 29475-64-7 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...