L-Alanine benzyl ester hydrochloride 98%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Ala-OBzl.HCl, H-Ala-OBzl·HCl

H65699-06 H65699-14 H65699-22
AAH65699-06EA 39.9 USD
AAH65699-06 AAH65699-14 AAH65699-22
L-Alanine benzyl ester hydrochloride 98%
L-Alanine benzyl ester hydrochloride

L-Alanine benzyl ester hydrochloride, 98%


Purity: 98% CAS No.: 5557-83-5 Molecular Formula: C10H13NO2.HCl Formula Weight: 215.68 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White to off-white MDL No.: MFCD00054340

Formula: C₁₀H₁₃NO₂
MW: 215.68 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00054340
CAS Number: 5557-83-5
EINECS: 226-920-4

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR