N-Me-Val-OMe·HCl 95%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Me-Val-OMe HCl, N-Methyl-L-valine methyl ester hydrochloride, N-Me-Val-OMe·HCl

H65254-03 H65254-MD
AAH65254-03EA 189 USD
AAH65254-03 AAH65254-MD
N-Me-Val-OMe·HCl 95%
N-Me-Val-OMe·HCl

N-Methyl-L-valine methyl ester hydrochloride, 95%


Purity: 95% CAS No.: 3339-44-4 Molecular Formula: C7H15NO2.HCl Formula Weight: 181.66 Storage Temperature: 2°C to 8°C Physical Form: Solid Melting Point: 103-104° Beilstein Reference: 3562296 MDL No.: MFCD00238325

Formula: C₇H₁₆ClNO₂
MW: 181.66 g/mol
Melting Pt: 103…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00238325
CAS Number: 3339-44-4

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR