Methoxyacetic anhydride 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Methoxyacetic Anhydride
H64541-14
H64541-22
AAH64541-14EA
108
USD
AAH64541-14
AAH64541-22
Methoxyacetic anhydride 95%
Methoxyacetic anhydride
Methoxyacetic anhydride, 95%
Purity: 95%
CAS No.: 19500-95-9
Molecular Formula: C6H10O5
Formula Weight: 162.14
Physical Form: Liquid
Sensitivity: Moisture Sensitive
Appearance: Colorless
Beilstein Reference: 1771552
UN No.: 3265
MDL No.: MFCD11976032
Formula:
C₆H₁₀O₅ MW: 162.141 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11976032 CAS Number: 19500-95-9 EINECS: 243-114-8 UN: 3265 ADR: 8 III |
Specification Test Results
Assay Minimum | 95 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...