2-[4-(Trifluoromethyl)phenyl]ethylamine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-(4-Trifluoromethylphenyl)ethylamine, 4-(Trifluoromethyl)phenethylamine
H64415-03
H64415-06
H64415-MD
AAH64415-03EA
64.6
USD
AAH64415-03
AAH64415-06
AAH64415-MD
2-[4-(Trifluoromethyl)phenyl]ethylamine 98%
2-[4-(Trifluoromethyl)phenyl]ethylamine
2-[4-(Trifluoromethyl)phenyl]ethylamine, 98%
Purity: 98%
CAS No.: 775-00-8
Molecular Formula: C9H10F3N
Formula Weight: 189.18
Beilstein Reference: 2719818
UN No.: 2735
MDL No.: MFCD01529875
Formula:
C₉H₁₀F₃N MW: 189.181 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01529875 CAS Number: 775-00-8 UN: 2735 ADR: "8",III |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...