9-(4-Bromophenyl)carbazole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
9-(4-Bromophenyl)-9H-carbazole
H64404-03
H64404-06
AAH64404-03EA
56.3
USD
AAH64404-03
AAH64404-06
AAH64404-14
9-(4-Bromophenyl)carbazole 98%
9-(4-Bromophenyl)carbazole
9-(4-Bromophenyl)carbazole, 98%
Purity: 98%
CAS No.: 57102-42-8
Molecular Formula: C18H12BrN
Formula Weight: 322.20
Physical Form: Solid
Melting Point: 147-151°
Beilstein Reference: 199590
MDL No.: MFCD11617969
Formula:
C₁₈H₁₂BrN MW: 322.21 g/mol Melting Pt: 147…151 °C Storage Temperature: Ambient |
MDL Number:
MFCD11617969 CAS Number: 57102-42-8 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...