2,4-Dichloro-6-methylpyridine 97+%

Supplier: Thermo Scientific Chemicals
Warning

H64248-03 H64248-06
AAH64248-03EA 76.5 USD
AAH64248-03 AAH64248-06 AAH64248-MD
2,4-Dichloro-6-methylpyridine 97+%
2,4-Dichloro-6-methylpyridine
Formula: C₆H₅Cl₂N
Boiling Pt: 205…206 °C
Storage Temperature: Ambient
MDL Number: MFCD08669834
CAS Number: 42779-56-6

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR