4-Formylbenzeneboronic acid pinacol ester 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Formylphenylboronic acid pinacol ester, 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde, 4-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
H64192-03
H64192-06
H64192-14
AAH64192-03EA
35.8
USD
AAH64192-03
AAH64192-06
AAH64192-14
4-Formylbenzeneboronic acid pinacol ester 98%
4-Formylbenzeneboronic acid pinacol ester
4-Formylbenzeneboronic acid pinacol ester, 98%
Purity: 98%
CAS No.: 128376-64-7
Molecular Formula: C13H17BO3
Formula Weight: 232.09
Physical Form: Crystalline solid
Appearance: Off-white
Melting Point: 54-59°
Beilstein Reference: 7704448
MDL No.: MFCD04972375
Formula:
C₁₃H₁₇BO₃ MW: 232.086 g/mol Melting Pt: 54…59 °C Storage Temperature: Ambient |
MDL Number:
MFCD04972375 CAS Number: 128376-64-7 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...