2-Cyano-6-methylpyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
6-Methylpicolinonitrile, 6-Methyl-2-pyridinecarbonitrile, 6-Cyano-2-picoline, 6-Methylpyridine-2-carbonitrile
H64148-06
H64148-14
AAH64148-06EA
47.3
USD
AAH64148-06
AAH64148-14
2-Cyano-6-methylpyridine 98%
2-Cyano-6-methylpyridine
2-Cyano-6-methylpyridine, 98%
Purity: 98%
CAS No.: 1620-75-3
Molecular Formula: C7H6N2
Formula Weight: 118.14
Physical Form: Solid
Appearance: Off-white
Melting Point: 70-74°
Beilstein Reference: 111107
UN No.: 3439
MDL No.: MFCD00190586
Formula:
C₇H₆N₂ MW: 118.14 g/mol Melting Pt: 70…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00190586 CAS Number: 1620-75-3 EINECS: 216-588-9 ADR: 6.1,III |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...