5-Bromo-2-chloroaniline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64104-06
H64104-14
H64104-22
AAH64104-06EA
43.9
USD
AAH64104-06
AAH64104-14
AAH64104-22
5-Bromo-2-chloroaniline 98%
5-Bromo-2-chloroaniline
5-Bromo-2-chloroaniline, 98%
Purity: 98%
CAS No.: 60811-17-8
Molecular Formula: C6H5BrClN
Formula Weight: 206.47
Physical Form: Solid
Melting Point: 145-150°
Beilstein Reference: 2716274
UN No.: 2811
MDL No.: MFCD06796283
Formula:
C₆H₅BrClN MW: 206.47 g/mol Melting Pt: 145…150 °C Storage Temperature: Ambient |
MDL Number:
MFCD06796283 CAS Number: 60811-17-8 UN: 2811 ADR: "6.1",III |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...