2-Chloro-6-methoxy-3-nitropyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64076-14
H64076-22
AAH64076-14EA
78.6
USD
AAH64076-14
AAH64076-22
2-Chloro-6-methoxy-3-nitropyridine 98%
2-Chloro-6-methoxy-3-nitropyridine
2-Chloro-6-methoxy-3-nitropyridine, 98%
Purity: 98%
CAS No.: 38533-61-8
Molecular Formula: C6H5ClN2O3
Formula Weight: 188.57
Physical Form: Powder
Appearance: Off-white to yellow
Melting Point: 78-80° lit.
Beilstein Reference: 1454907
UN No.: 2811
MDL No.: MFCD00130268
Formula:
C₆H₅ClN₂O₃ Melting Pt: 78…80 °C (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00130268 CAS Number: 38533-61-8 EINECS: 253-989-8 ADR: 6.1,III |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...