2-Bromo-5-chloro-3-nitropyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

H64065-06 H64065-14
AAH64065-03EA 22.98 USD
AAH64065-03 AAH64065-06 AAH64065-14
2-Bromo-5-chloro-3-nitropyridine 98%
2-Bromo-5-chloro-3-nitropyridine

2-Bromo-5-chloro-3-nitropyridine, 98%


Purity: 98% CAS No.: 75806-86-9 Molecular Formula: C5H2BrClN2O2 Formula Weight: 237.44 Physical Form: Solid Appearance: Off-white Melting Point: 175-179° Beilstein Reference: 147540 UN No.: 2811 MDL No.: MFCD07368982

Formula: C₅H₂BrClN₂O₂
Melting Pt: 175…179 °C
Storage Temperature: Ambient
MDL Number: MFCD07368982
CAS Number: 75806-86-9
UN: 2811
ADR: "6.1",III

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR