3-Methyl-3-(p-toluenesulfonyloxymethyl)oxetane 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (3-Methyloxetan-3-yl)methyl 4-methylbenzenesulphonate

H64011-03 H64011-06
AAH64011-03EA 104 USD
AAH64011-03 AAH64011-06 AAH64011-MD
3-Methyl-3-(p-toluenesulfonyloxymethyl)oxetane 98%
3-Methyl-3-(p-toluenesulfonyloxymethyl)oxetane

3-Methyl-3-(p-toluenesulfonyloxymethyl)oxetane, 98%


Purity: 98% CAS No.: 99314-44-0 Molecular Formula: C12H16O4S Formula Weight: 256.32 Storage Temperature: 2°C to 8°C Physical Form: Solid Appearance: Off-white Melting Point: 50-51° Beilstein Reference: 4441465 MDL No.: MFCD08275394

Formula: C₁₂H₁₆O₄S
Melting Pt: 50…51 °C
Storage Temperature: Ambient
MDL Number: MFCD08275394
CAS Number: 99314-44-0

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR