1-Boc-indoline-7-carboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63883-03
H63883-06
AAH63883-03EA
64.9
USD
AAH63883-03
AAH63883-06
AAH63883-MD
1-Boc-indoline-7-carboxylic acid 97%
1-Boc-indoline-7-carboxylic acid
1-Boc-indoline-7-carboxylic acid, 97%
Purity: 97%
CAS No.: 143262-20-8
Molecular Formula: C14H17NO4
Formula Weight: 263.29
Physical Form: Solid
Sensitivity: Light Sensitive
Appearance: Pale Yellow
Melting Point: 182° dec.
UN No.: 3077
MDL No.: MFCD04973983
Formula:
C₁₄H₁₇NO₄ MW: 263.293 g/mol Melting Pt: 182 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD04973983 CAS Number: 143262-20-8 UN: 3077 ADR: "9",III |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...