DL-Phenylalaninol 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Amino-3-phenyl-1-propanol, DL-2-Amino-3-phenyl-1-propanol, 2-Amino-3-phenylpropan-1-ol

H63562-03 H63562-06 H63562-14
AAH63562-03EA 42.1 USD
AAH63562-03 AAH63562-06 AAH63562-14
DL-Phenylalaninol 96%
DL-Phenylalaninol

DL-Phenylalaninol, 96%


Purity: 96% CAS No.: 16088-07-6 Molecular Formula: C9H13NO Formula Weight: 151.21 Storage Temperature: -30°C to -10°C Physical Form: Solid Melting Point: 69-73° Beilstein Reference: 2208237 UN No.: 3259 MDL No.: MFCD00066689

Formula: C₉H₁₃NO
MW: 151.21 g/mol
Melting Pt: 69…73 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00066689
CAS Number: 16088-07-6
UN: 3259
ADR: "8",III

Order Now

Specification Test Results

Assay Minimum 96

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR