Phenol, 4-[(1S)-1-aminoethyl]- 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(S)-4-(1-Aminoethyl)phenol
H63269-03
H63269-06
AAH63269-03EA
272
USD
AAH63269-03
AAH63269-06
AAH63269-MD
Phenol, 4-[(1S)-1-aminoethyl]- 97%
Phenol, 4-[(1S)-1-aminoethyl]-
(S)-4-(1-Aminoethyl)phenol, 97%
Purity: 97%
CAS No.: 221670-72-0
Molecular Formula: C8H11NO
Formula Weight: 137.18
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: Off-white
UN No.: 3259
MDL No.: MFCD03844646
Formula:
C₈H₁₁NO Storage Temperature: Ambient |
MDL Number:
MFCD03844646 CAS Number: 221670-72-0 UN: 3259 ADR: "8",III |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...