2-Cyano-5-fluorobenzyl bromide 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Bromomethyl-4-fluorobenzonitrile

H63110-06 H63110-14
AAH63110-03EA 24.51 USD
AAH63110-03 AAH63110-06 AAH63110-14
2-Cyano-5-fluorobenzyl bromide 98%
2-Cyano-5-fluorobenzyl bromide

2-Bromomethyl-4-fluorobenzonitrile, 98%


Purity: 98% CAS No.: 421552-12-7 Molecular Formula: C8H5BrFN Formula Weight: 214.04 Physical Form: Solid UN No.: 2923 MDL No.: MFCD08059542

Formula: C8H5BrFN
MW: 214.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08059542
CAS Number: 421552-12-7
UN: 2923
ADR: "8",III

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR