O-Methyl-L-tyrosine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(S)-2-Amino-3-(4-methoxyphenyl)propionic Acid, H-Tyr(Me)-OH, 4-Methoxy-L-phenylalanine
H63096-03
H63096-06
AAH63096-03EA
34.5
USD
AAH63096-03
AAH63096-06
O-Methyl-L-tyrosine 98%
O-Methyl-L-tyrosine
O-Methyl-L-tyrosine, 98%
Purity: 98%
CAS No.: 6230-11-1
Molecular Formula: C10H13NO3
Formula Weight: 195.22
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 259-261° dec
Beilstein Reference: 2212726
Optical Rotation: -7°(c=1 in 1M HCl)
MDL No.: MFCD00002604
Formula:
C₁₀H₁₃NO₃ MW: 195.22 g/mol Melting Pt: 259…261 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00002604 CAS Number: 6230-11-1 EINECS: 228-333-9 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...