3-Fluoro-4-(4-morpholinylmethyl)benzeneboronic acid pinacol ester 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)morpholine

H62530-03 H62530-MD
AAH62530-03EA 214 USD
AAH62530-03 AAH62530-MD
3-Fluoro-4-(4-morpholinylmethyl)benzeneboronic acid pinacol ester 96%
3-Fluoro-4-(4-morpholinylmethyl)benzeneboronic acid pinacol ester

3-Fluoro-4-(4-morpholinylmethyl)benzeneboronic acid pinacol ester, 96%


Purity: 96% CAS No.: 1073354-74-1 Molecular Formula: C17H25BFNO3 Formula Weight: 321.19 Physical Form: Solid Melting Point: 35-40° MDL No.: MFCD09027076

Formula: C₁₇H₂₅BFNO₃
Melting Pt: 35…40 °C
Storage Temperature: Ambient
MDL Number: MFCD09027076
CAS Number: 1073354-74-1

Order Now

Specification Test Results

Assay Minimum 96


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR