Boc-N-Me-Phe-OH 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Boc-N-methyl-L-phenylalanine
H62221-03
H62221-06
AAH62221-03EA
55.7
USD
AAH62221-03
AAH62221-06
Boc-N-Me-Phe-OH 95%
Boc-N-Me-Phe-OH
N-Boc-N-methyl-L-phenylalanine, 95%
Purity: 95%
CAS No.: 37553-65-4
Molecular Formula: C15H21NO4
Formula Weight: 279.33
Physical Form: Solid
Beilstein Reference: 2386503
MDL No.: MFCD00038754
Formula:
C₁₅H₂₁NO₄ MW: 279.336 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00038754 CAS Number: 37553-65-4 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...