p-Toluenesulfonylsemicarbazide 95%

Supplier: Thermo Scientific Chemicals

H61949-14 H61949-22
AAH61949-14EA 72.7 USD
AAH61949-14 AAH61949-22
p-Toluenesulfonylsemicarbazide 95%
p-Toluenesulfonylsemicarbazide

p-Toluenesulfonyl semicarbazide, 95%


Purity: 95% CAS No.: 10396-10-8 Molecular Formula: C8H11N3O3S Formula Weight: 229.26 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White MDL No.: MFCD00072243

Formula: C8H11N3O3S
MW: 229.26 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00072243
CAS Number: 10396-10-8
EINECS: 233-857-6

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR