2-Bromo-3,4-difluoroaniline 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H61876-06
H61876-14
AAH61876-03EA
40.06
USD
AAH61876-03
AAH61876-06
AAH61876-14
2-Bromo-3,4-difluoroaniline 97%
2-Bromo-3,4-difluoroaniline
2-Bromo-3,4-difluoroaniline, 97%
Purity: 97%
CAS No.: 1092349-87-5
Molecular Formula: C6H4BrF2N
Formula Weight: 208.00
Physical Form: Solid
UN No.: 2811
MDL No.: MFCD11035924
Formula:
C₆H₄BrF₂N MW: 208.00 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11035924 CAS Number: 1092349-87-5 UN: 2811 ADR: "6.1",III |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...