4,6-Dichloro-5-fluoropyrimidine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H61770-03
H61770-06
AAH61770-03EA
156
USD
AAH61770-03
AAH61770-06
4,6-Dichloro-5-fluoropyrimidine 98%
4,6-Dichloro-5-fluoropyrimidine
4,6-Dichloro-5-fluoropyrimidine, 98%
Purity: 98%
CAS No.: 213265-83-9
Molecular Formula: C4HCl2FN2
Formula Weight: 166.97
Flash Point: 91°(196°F)
Physical Form: Liquid
Boiling Point: 50-60°/14mm
UN No.: 3265
MDL No.: MFCD08056331
Formula:
C₄HCl₂FN₂ MW: 166.97 g/mol Boiling Pt: 50…60 °C/14 mm Flash Pt: 91 °C (196 °F) Storage Temperature: Ambient |
MDL Number:
MFCD08056331 CAS Number: 213265-83-9 UN: 3265 ADR: 8,III |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...