5-Chlorothiophene-2-carbonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

H61474-06 H61474-14
AAH61474-06EA 128 USD
AAH61474-06 AAH61474-14
5-Chlorothiophene-2-carbonyl chloride 98%
5-Chlorothiophene-2-carbonyl chloride

5-Chlorothiophene-2-carbonyl chloride, 98%


Purity: 98% CAS No.: 42518-98-9 Molecular Formula: C5H2Cl2OS Formula Weight: 181.04 Storage Temperature: 2°C to 8°C Melting Point: 4° UN No.: 3265 MDL No.: MFCD00043886

Formula: C5H2Cl2OS
MW: 181.04 g/mol
Melting Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00043886
CAS Number: 42518-98-9
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR