Hematoxylin hydrate, (max. 8% H₂O) ≥80% (dry weight), powder
Supplier: Thermo Scientific Chemicals
|
Warning
|
047223-14
047223-22
AA47223-06EA
33.89
USD
AA47223-06
AA47223-14
AA47223-22
Hematoxylin hydrate, (max. 8% H₂O) ≥80% (dry weight), powder
Hematoxylin hydrate
Hematoxylin hydrate, 80% (dry wt.), water <8%. Cream to brown powder.
Purity: 80%
CAS No.: 517-28-2
Molecular Formula: C16H14O6.xH2O
Formula Weight: 302.29(anhy)
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: Cream to brown
Melting Point: ca 200° dec.
Beilstein Reference: 91399
MDL No.: MFCD00078111
Formula:
C₁₆H₁₄O₆·xH₂O Melting Pt: ∼ 200 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00078111 CAS Number: 517-28-2 EINECS: 208-237-3 Merck Index: 14,4637 |
Specification Test Results
Assay Minimum | 80 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...