Scandium(III) bromide, ultra dry ≥99.99% (metals basis), powder
Supplier: Thermo Scientific Chemicals
ampouled under argon
Scandium(III) bromide, ultra dry, 99.99% (metals basis). Powder, Ampouled under argon.
Purity: 99.99%
CAS No.: 13465-59-3
Molecular Formula: ScBr3
Formula Weight: 284.67
Physical Form: Powder, Ampouled under argon
Sensitivity: Hygroscopic
MDL No.: MFCD00016324
Formula:
Br₃Sc Storage Temperature: Ambient |
MDL Number:
MFCD00016324 CAS Number: 13465-59-3 EINECS: 236-699-6 |
Specification Test Results
Assay Minimum | 99.99 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...