Thulium(III) bromide, ultra dry ≥99.99% (REO, rare earth oxide basis), beads
Supplier: Thermo Scientific Chemicals
|
Warning
|
ampouled under argon
047196-03
047196-06
AA47196-03EA
189
USD
AA47196-03
AA47196-06
Thulium(III) bromide, ultra dry ≥99.99% (REO, rare earth oxide basis), beads
Thulium(III) bromide
Thulium(III) bromide, ultra dry, 99.99% (REO). -10 Mesh Beads, Ampouled under argon.
Purity: 99.99%
CAS No.: 14456-51-0
Molecular Formula: TmBr3
Formula Weight: 408.66
Physical Form: -10 Mesh Beads, Ampouled under argon
Sensitivity: Hygroscopic
Melting Point: 952°
Boiling Point: 1440°
MDL No.: MFCD00050138
Formula:
Br₃Tm MW: 408.65 g/mol Boiling Pt: 1440 °C Melting Pt: 952 °C Storage Temperature: Ambient |
MDL Number:
MFCD00050138 CAS Number: 14456-51-0 EINECS: 238-444-4 |
Specification Test Results
Assay Minimum | 99.99 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...